Merck
  • Activity-dependent post-translational regulation of palmitoylating and depalmitoylating enzymes in the hippocampus.
Activity-dependent post-translational regulation of palmitoylating and depalmitoylating enzymes in the hippocampus.
Journal of Cell Science (2023)
Abazari, et al.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Millipore
ANTI-FLAG® in Kaninchen hergestellte Antikörper, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-β-Actin-Antikörper, Maus monoklonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-ZDHHC5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-ZDHHC9 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-ZDHHC17 (AB1) antibody produced in rabbit, affinity isolated antibody