Merck
  • Biallelic mutations in RNA-binding protein ADAD2 cause spermiogenic failure and non-obstructive azoospermia in humans.
Biallelic mutations in RNA-binding protein ADAD2 cause spermiogenic failure and non-obstructive azoospermia in humans.
Human reproduction open (2023-06-16)
Baolu Shi, Wasim Shah, Li Liu, Chenjia Gong, Jianteng Zhou, Tanveer Abbas, Hui Ma, Huan Zhang, Menglei Yang, Yuanwei Zhang, Nadeem Ullah, Zubair Mahammad, Mazhar Khan, Ghulam Murtaza, Asim Ali, Ranjha Khan, Jiahao Sha, Yan Yuan, Qinghua Shi
ZUSAMMENFASSUNG
What are some pathogenic mutations for non-obstructive azoospermia (NOA) and their effects on spermatogenesis? Biallelic missense and frameshift mutations in ADAD2 disrupt the differentiation of round spermatids to spermatozoa causing azoospermia in humans and mice. NOA is the most severe cause of male infertility characterized by an absence of sperm in the ejaculate due to impairment of spermatogenesis. In mice, the lack of the RNA-binding protein ADAD2 leads to a complete absence of sperm in epididymides due to failure of spemiogenesis, but the spermatogenic effects of ADAD2 mutations in human NOA-associated infertility require functional verification. Six infertile male patients from three unrelated families were diagnosed with NOA at local hospitals in Pakistan based on infertility history, sex hormone levels, two semen analyses and scrotal ultrasound. Testicular biopsies were performed in two of the six patients. Adad2 mutant mice (Adad2Mut/Mut) carrying mutations similar to those found in NOA patients were generated using the CRISPR/Cas9 genome editing tool. Reproductive phenotypes of Adad2Mut/Mut mice were verified at 2 months of age. Round spermatids from the littermates of wild-type (WT) and Adad2Mut/Mut mice were randomly selected and injected into stimulated WT oocytes. This round spermatid injection (ROSI) procedure was conducted with three biological replicates and >400 ROSI-derived zygotes were evaluated. The fertility of the ROSI-derived progeny was evaluated for three months in four Adad2WT/Mut male mice and six Adad2WT/Mut female mice. A total of 120 Adad2Mut/Mut, Adad2WT/Mut, and WT mice were used in this study. The entire study was conducted over 3 years. Whole-exome sequencing was performed to detect potentially pathogenic mutations in the six NOA-affected patients. The pathogenicity of the identified ADAD2 mutations was assessed and validated in human testicular tissues and in mouse models recapitulating the mutations in the NOA patients using quantitative PCR, western blotting, hematoxylin-eosin staining, Periodic acid-Schiff staining, and immunofluorescence. Round spermatids of WT and Adad2Mut/Mut mice were collected by fluorescence-activated cell sorting and injected into stimulated WT oocytes. The development of ROSI-derived offspring was evaluated in the embryonic and postnatal stages. Three recessive mutations were identified in ADAD2 (MT1: c.G829T, p.G277C; MT2: c.G1192A, p.D398N; MT3: c.917_918del, p.Q306Rfs*43) in patients from three unrelated Pakistani families. MT1 and MT2 dramatically reduced the testicular expression of ADAD2, likely causing spermiogenesis failure in the NOA patients. Immunofluorescence analysis of the Adad2Mut/Mut male mice with the corresponding MT3 mutation showed instability and premature degradation of the ADAD2 protein, resulting in the spermiogenesis deficiency phenotype. Through ROSI, the Adad2Mut/Mut mice could produce pups with comparable embryonic development (46.7% in Adad2Mut/Mut versus 50% in WT) and birth rates (21.45 ± 10.43% in Adad2Mut/Mut versus 27.5 ± 3.536% in WT, P = 0.5044) to WT mice. The Adad2WT/Mut progeny from ROSI (17 pups in total via three ROSI replicates) did not show overt developmental defects and had normal fertility. N/A. This is a preliminary report suggesting that ROSI can be an effective treatment for infertile Adad2Mut/Mut mice. Further assisted reproductive attempts need to be carefully examined in humans during clinical trials. Our work provides functional evidence that mutations in the ADAD2 gene are deleterious and cause consistent spermiogenic defects in both humans and mice. In addition, preliminary results show that ROSI can help Adad2Mut/Mut to produce biological progeny. These findings provide valuable clues for genetic counselling on the ADAD2 mutants-associated infertility in human males. This work was supported by the National Natural Science Foundation of China (32000587, U21A20204, and 32061143006), and the National Key Research and Developmental Program of China (2019YFA0802600 and 2021YFC2700202). This work was also supported by Institute of Health and Medicine, Hefei Comprehensive National Science Center, Hefei, China. The authors declare no competing interests.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cytochalasin B aus Drechslera dematioidea, ≥98% (HPLC), powder
Sigma-Aldrich
Hyaluronidase aus Rindertestes, Type IV-S, powder, suitable for mouse embryo cell culture, 750-3000 units/mg solid
Sigma-Aldrich
Collagenase Typ IV, Cls IV