Merck
Iron-loaded deferiprone can support full hemoglobinization of cultured red blood cells.
Scientific Reports (2023)
Gallego-Murillo, et al.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler Anti-Aktin-Antikörper der Maus, clone AC-40, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Phospho-STAT5A/B(Tyr694/699)-Antikörper, Klon 8-5-2, clone 8-5-2, Upstate®, from mouse
Sigma-Aldrich
Anti-TFRC antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody