Merck
  • The Impact of In Vitro Digestion on the Polyphenol Content and Antioxidant Activity of Spanish Ciders.
The Impact of In Vitro Digestion on the Polyphenol Content and Antioxidant Activity of Spanish Ciders.
Foods (Basel, Switzerland) (2023-05-13)
Mari Mar Cavia, Nerea Arlanzón, Natalia Busto, Celia Carrillo, Sara R Alonso-Torre
ZUSAMMENFASSUNG
Various factors can influence the polyphenol content and the antioxidant capacity of ciders, such as the apple variety, its degree of maturity, apple farming and storage conditions, and the cider-fermentation method, all of which explains why ciders of different origin present different values. In addition, digestive processes could have some effects on the properties of cider. Hence, the objective of this study is to characterize Spanish ciders in terms of their polyphenol content and antioxidant capacity and to ascertain whether those same properties differ in digested ciders. In total, 19 ciders were studied from three different zones within Spain: Asturias (A) (10), the Basque Country (BC) (6), and Castile-and-Leon (CL) (3). A range of assays was used to determine the total polyphenol content and the antioxidant capacity of the ciders. In addition, a digestive process was simulated in vitro, assessing whether the use of amylase might influence the recovery of bioactive compounds after digestion. The Basque Country ciders presented higher total polyphenol contents (830 ± 179 GAE/L) and higher antioxidant capacities (DPPH: 5.4 ± 1.6 mmol TE/L; ABTS: 6.5 ± 2.0 mmol TE/L; FRAP: 6.9 ± 1.6 mmol TE/L) than the other ciders that were studied. The in vitro digestion process, regardless of the use of amylase, implied a loss of phenolic compounds (598 ± 239 mg GAE/L undigested samples; 466 ± 146 mg GAE/L digested without amylase samples; 420 ± 115 mg GAE/L digested with amylase samples), although the variation in antioxidant activity depended on the assay chosen for its determination.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pancreatin aus Schweinepankreas, 4 × USP specifications