Merck
  • Proliferating cell nuclear antigen impairs the nuclear import of influenza A virus PB2 and suppresses virus replication.
Proliferating cell nuclear antigen impairs the nuclear import of influenza A virus PB2 and suppresses virus replication.
Journal of medical virology (2023-06-07)
Yuanbao Zhou, Shishuo Zhang, Biaoxiong Wang, Ying Ran, Shaoyu Tu, Zhipeng Lin, Meilin Jin, Huanchun Chen, Jiahui Zou, Hongbo Zhou
ZUSAMMENFASSUNG
The genome of Influenza A virus (IAV) transcribes and replicates in the nucleus of cells and the viral ribonucleoprotein (vRNP) complex plays an important role in viral replication. As a major component of the vRNP complex, the polymerase basic protein 2 (PB2) is translocated to the nucleus via its nuclear localization signals mediated by the importins. Herein, it was identified proliferating cell nuclear antigen (PCNA) as an inhibitor of nuclear import of PB2 and subsequent viral replication. Mechanically, PCNA interacted with PB2 and inhibited the nuclear import of PB2. Furthermore, PCNA decreased the binding efficiency of PB2 with importin alpha (importin α) and the K738, K752, and R755 of PB2 were identified as the key sites binding with PCNA and importin α. Furthermore, PCNA was demonstrated to retrain the vRNP assembly and polymerase activity. Taken together, the results demonstrated that PCNA impaired the nuclear import of PB2, vRNP assembly and polymerase activity, which negatively regulated virus replication.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)