Merck
  • The disulfide catalyst QSOX1 maintains the colon mucosal barrier by regulating Golgi glycosyltransferases.
The disulfide catalyst QSOX1 maintains the colon mucosal barrier by regulating Golgi glycosyltransferases.
The EMBO journal (2022-10-18)
Tal Ilani, Nava Reznik, Noa Yeshaya, Tal Feldman, Patrick Vilela, Zipora Lansky, Gabriel Javitt, Michal Shemesh, Ori Brenner, Yoav Elkis, Neta Varsano, Ana M Jaramillo, Christopher M Evans, Deborah Fass
ZUSAMMENFASSUNG
Mucus is made of enormous mucin glycoproteins that polymerize by disulfide crosslinking in the Golgi apparatus. QSOX1 is a catalyst of disulfide bond formation localized to the Golgi. Both QSOX1 and mucins are highly expressed in goblet cells of mucosal tissues, leading to the hypothesis that QSOX1 catalyzes disulfide-mediated mucin polymerization. We found that knockout mice lacking QSOX1 had impaired mucus barrier function due to production of defective mucus. However, an investigation on the molecular level revealed normal disulfide-mediated polymerization of mucins and related glycoproteins. Instead, we detected a drastic decrease in sialic acid in the gut mucus glycome of the QSOX1 knockout mice, leading to the discovery that QSOX1 forms regulatory disulfides in Golgi glycosyltransferases. Sialylation defects in the colon are known to cause colitis in humans. Here we show that QSOX1 redox control of sialylation is essential for maintaining mucosal function.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Roche
DAPI, 4′,6-Diamidine-2′-phenylindole dihydrochloride
Sigma-Aldrich
Anti-ST6GAL1 antibody produced in rabbit, affinity isolated antibody