Merck
  • FBXL6 is dysregulated in keloids and promotes keloid fibroblast growth by inducing c-Myc expression.
FBXL6 is dysregulated in keloids and promotes keloid fibroblast growth by inducing c-Myc expression.
International wound journal (2022-05-24)
Guangdong Feng, Hui Sun, Meishan Piao
ZUSAMMENFASSUNG
C-MYC-mediated keloid fibroblasts proliferation and collagen deposit may contribute to the development of keloids. F-box and leucine-rich repeat protein 6 (FBXL6) is reported to be involved in tumour progression, while the role of FBXL6 in keloid fibroblasts is not deciphered. Normal control skins, hypertrophic scars and keloid tissues were collected and prepared for FBXL6 detection. FBXL6 short hairpin RNAs (shRNAs) or FBXL6 over-expression plasmids were transfected into keloid fibroblasts, and then c-MYC plasmids were further transfected. Cell viability was assayed with a Cell-Counting Kit-8 kit. The relative expression of FBXL6, Cyclin A1, Cyclin D2, Cyclin E1 and Collagen I was detected with real-time PCR and Western blot. Elevated FBXL6 expression could be observed in keloid tissues and hypertrophic scars. FBXL6 shRNAs transfection could inhibit the viability of keloid fibroblasts with diminished c-MYC expression and down-regulated Cyclin A1, Cyclin D2, Cyclin E1 and Collagen I expression. At the same time, overexpressed FBXL6 could promote the proliferation of keloid fibroblasts. Overexpression of c-MYC could promote the proliferation of keloid fibroblasts reduced by FBXL6 shRNAs with up-regulated Cyclin A1 and Collagen I expression. FBXL6 could promote the growth of keloid fibroblasts by inducing c-MYC expression, which could be targeted in keloids treatment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-FBXL6 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution