Merck
  • Glycine Alleviated Intestinal Injury by Inhibiting Ferroptosis in Piglets Challenged with Diquat.
Glycine Alleviated Intestinal Injury by Inhibiting Ferroptosis in Piglets Challenged with Diquat.
Animals : an open access journal from MDPI (2022-11-27)
Xiao Xu, Yu Wei, Hongwei Hua, Huiling Zhu, Kan Xiao, Jiangchao Zhao, Yulan Liu
ZUSAMMENFASSUNG
The purpose of this research was to examine the impact of glycine on intestinal injury caused by oxidative stress in piglets. A 2 × 2 factorial experiment with diets (basic diet vs. 1% glycine diet) and oxidative stress (saline vs. diquat) was conducted on 32 weanling piglets. On day 21, all piglets received an injection of either saline or diquat. After 7 days, all pigs were slaughtered and intestinal samples were collected. Dietary glycine supplementation improved intestinal mucosal morphology, increased the activities of disaccharidases and enhanced intestinal mucosal antioxidant capacity, while regulating the expression of ferroptosis mediators in the piglets under oxidative stress. These findings suggested that dietary glycine supplementation improved the morphology and function of the intestinal mucosa, which was involved in regulating antioxidant capacity and ferroptosis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-74, purified immunoglobulin, buffered aqueous solution