Merck
FLT4 as a marker for predicting prognostic risk of refractory acute myeloid leukemia.
Haematologica (2023-06-15)
Ji Yoon Lee, Sung-Eun Lee, A-Reum Han, Jongeun Lee, Young-Sup Yoon, Hee-Je Kim
ZUSAMMENFASSUNG
Treating patients with refractory acute myeloid leukemia (AML) remains challenging. Currently there is no effective treatment for refractory AML. Increasing evidence has demonstrated that refractory/relapsed AML is associated with leukemic blasts which can confer resistance to anticancer drugs. We have previously reported that high expression of Fms-related tyrosine kinase 4 (FLT4) is associated with increased cancer activity in AML. However, the functional role of FLT4 in leukemic blasts remains unknown. Here, we explored the significance of FLT4 expression in leukemic blasts of refractory patients and mechanisms involved in the survival of AML blasts. Inhibition or absence of FLT4 in AML-blasts suppressed homing to bone marrow (BM) of immunocompromised mice and blocked engraftment of AML blasts. Moreover, FLT4 inhibition by MAZ51, an antagonist, effectively reduced the number of leukemic cell-derived colonyforming units and increased apoptosis of blasts derived from refractory patients when it was cotreated with cytosine arabinoside (Ara-C) under VEGF-C, its ligand. AML patients who expressed high cytosolic FLT4 were linked to an AML-refractory status by internalization mechanism. In conclusion, FLT4 has a biological function in leukemogenesis and refractoriness. This novel insight will be useful for targeted therapy and prognostic stratification of AML.