Merck
PCNA recruits cohesin loader Scc2 to ensure sister chromatid cohesion.
Nature structural & molecular biology (2023-08-18)
Ivan Psakhye, Ryotaro Kawasumi, Takuya Abe, Kouji Hirota, Dana Branzei
ZUSAMMENFASSUNG
Sister chromatid cohesion, established during replication by the ring-shaped multiprotein complex cohesin, is essential for faithful chromosome segregation. Replisome-associated proteins are required to generate cohesion by two independent pathways. One mediates conversion of cohesins bound to unreplicated DNA ahead of replication forks into cohesive entities behind them, while the second promotes cohesin de novo loading onto newly replicated DNA. The latter process depends on the cohesin loader Scc2 (NIPBL in vertebrates) and the alternative PCNA loader CTF18-RFC. However, the mechanism of de novo cohesin loading during replication is unknown. Here we show that PCNA physically recruits the yeast cohesin loader Scc2 via its C-terminal PCNA-interacting protein motif. Binding to PCNA is crucial, as the scc2-pip mutant deficient in Scc2-PCNA interaction is defective in cohesion when combined with replisome mutants of the cohesin conversion pathway. Importantly, the role of NIPBL recruitment to PCNA for cohesion generation is conserved in vertebrate cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Roche
Anti-HA-Biotin, High Affinity (3F10), from rat IgG1