Merck
PRMT5 facilitates angiogenesis and EMT via HIF-1α/VEGFR/Akt signaling axis in lung cancer.
Aging (2023-07-04)
Yonghua Zheng, Huaxia Ji, Wulin Yi, Zhanjun Chen, Xiaobiao Hu, Jie Zhou, Yang Wang, Xiao Zheng
ZUSAMMENFASSUNG
Abnormal angiogenesis is a critical factor in tumor growth and metastasis, and protein arginine methyltransferase 5 (PRMT5), a prominent type II enzyme, is implicated in various human cancers. However, the precise role of PRMT5 in regulating angiogenesis to promote lung cancer cell metastasis and the underlying molecular mechanisms are not fully understood. Here, we show that PRMT5 is overexpressed in lung cancer cells and tissues, and its expression is triggered by hypoxia. Moreover, inhibiting or silencing PRMT5 disrupts the phosphorylation of the VEGFR/Akt/eNOS angiogenic signaling pathway, NOS activity, and NO production. Additionally, inhibiting PRMT5 activity reduces HIF-1α expression and stability, resulting in the down-regulation of the VEGF/VEGFR signaling pathway. Our findings indicate that PRMT5 promotes lung cancer epithelial-mesenchymal transition (EMT), which might be possibly through controlling the HIF-1α/VEGFR/Akt/eNOS signaling axis. Our study provides compelling evidence of the close association between PRMT5 and angiogenesis/EMT and highlights the potential of targeting PRMT5 activity as a promising therapeutic approach for treating lung cancer with abnormal angiogenesis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fetales Kälberserum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
DAPI, for nucleic acid staining
Millipore
ANTI-FLAG® in Kaninchen hergestellte Antikörper, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Puromycin -dihydrochlorid, Ready Made Solution, from Streptomyces alboniger, 10 mg/mL in H2O, suitable for cell culture