Merck
  • AIM-CICs: an automatic identification method for cell-in-cell structures based on convolutional neural network.
AIM-CICs: an automatic identification method for cell-in-cell structures based on convolutional neural network.
Journal of molecular cell biology (2022-07-24)
Meng Tang, Yan Su, Wei Zhao, Zubiao Niu, Banzhan Ruan, Qinqin Li, You Zheng, Chenxi Wang, Bo Zhang, Fuxiang Zhou, Xiaoning Wang, Hongyan Huang, Hanping Shi, Qiang Sun
ZUSAMMENFASSUNG
Edited by Luonan Chen Whereas biochemical markers are available for most types of cell death, current studies on non-autonomous cell death by entosis rely strictly on the identification of cell-in-cell structures (CICs), a unique morphological readout that can only be quantified manually at present. Moreover, the manual CIC quantification is generally over-simplified as CIC counts, which represents a major hurdle against profound mechanistic investigations. In this study, we take advantage of artificial intelligence technology to develop an automatic identification method for CICs (AIM-CICs), which performs comprehensive CIC analysis in an automated and efficient way. The AIM-CICs, developed on the algorithm of convolutional neural network, can not only differentiate between CICs and non-CICs (the area under the receiver operating characteristic curve (AUC) > 0.99), but also accurately categorize CICs into five subclasses based on CIC stages and cell number involved (AUC > 0.97 for all subclasses). The application of AIM-CICs would systemically fuel research on CIC-mediated cell death, such as high-throughput screening.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4′,6-Diamidin-2-phenylindol -dihydrochlorid, powder, BioReagent, suitable for cell culture, ≥98% (HPLC), suitable for fluorescence