Merck
Human Dectin-1 is O-glycosylated and serves as a ligand for C-type lectin receptor CLEC-2.
eLife (2022-12-09)
Shojiro Haji, Taiki Ito, Carla Guenther, Miyako Nakano, Takashi Shimizu, Daiki Mori, Yasunori Chiba, Masato Tanaka, Sushil K Mishra, Janet A Willment, Gordon D Brown, Masamichi Nagae, Sho Yamasaki
ZUSAMMENFASSUNG
C-type lectin receptors (CLRs) elicit immune responses upon recognition of glycoconjugates present on pathogens and self-components. While Dectin-1 is the best-characterized CLR recognizing β-glucan on pathogens, the endogenous targets of Dectin-1 are not fully understood. Herein, we report that human Dectin-1 is a ligand for CLEC-2, another CLR expressed on platelets. Biochemical analyses revealed that Dectin-1 is a mucin-like protein as its stalk region is highly O-glycosylated. A sialylated core 1 glycan attached to the EDxxT motif of human Dectin-1, which is absent in mouse Dectin-1, provides a ligand moiety for CLEC-2. Strikingly, the expression of human Dectin-1 in mice rescued the lethality and lymphatic defect resulting from a deficiency of Podoplanin, a known CLEC-2 ligand. This finding is the first example of an innate immune receptor also functioning as a physiological ligand to regulate ontogeny upon glycosylation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Zymosan A aus Saccharomyces cerevisiae, for inducing inflamatory response