Merck
  • IL-12 sensing in neurons induces neuroprotective CNS tissue adaptation and attenuates neuroinflammation in mice.
IL-12 sensing in neurons induces neuroprotective CNS tissue adaptation and attenuates neuroinflammation in mice.
Nature neuroscience (2023-09-26)
Myrto Andreadou, Florian Ingelfinger, Donatella De Feo, Teresa L M Cramer, Selma Tuzlak, Ekaterina Friebel, Bettina Schreiner, Pascale Eede, Shirin Schneeberger, Maria Geesdorf, Frederike Ridder, Christina A Welsh, Laura Power, Daniel Kirschenbaum, Shiva K Tyagarajan, Melanie Greter, Frank L Heppner, Sarah Mundt, Burkhard Becher
ZUSAMMENFASSUNG
Interleukin-12 (IL-12) is a potent driver of type 1 immunity. Paradoxically, in autoimmune conditions, including of the CNS, IL-12 reduces inflammation. The underlying mechanism behind these opposing properties and the involved cellular players remain elusive. Here we map IL-12 receptor (IL-12R) expression to NK and T cells as well as neurons and oligodendrocytes. Conditionally ablating the IL-12R across these cell types in adult mice and assessing their susceptibility to experimental autoimmune encephalomyelitis revealed that the neuroprotective role of IL-12 is mediated by neuroectoderm-derived cells, specifically neurons, and not immune cells. In human brain tissue from donors with multiple sclerosis, we observe an IL-12R distribution comparable to mice, suggesting similar mechanisms in mice and humans. Combining flow cytometry, bulk and single-nucleus RNA sequencing, we reveal an IL-12-induced neuroprotective tissue adaption preventing early neurodegeneration and sustaining trophic factor release during neuroinflammation, thereby maintaining CNS integrity in mice.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Roche
cOmplete, Mini, EDTA-freier Protease-Inhibitor-Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Roche
cOmplete Mini Proteasehemmer-Cocktail, Tablets provided in a glass vial
Sigma-Aldrich
Phosphatasehemmer-Cocktail 3, DMSO solution
Sigma-Aldrich
Monoklonaler Anti-MAP2 in Maus hergestellte Antikörper, ascites fluid, clone HM-2
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 1, DMSO solution