Merck
Modifications of The Human Liver Cancer Cells through microRNA-145-Mediated Targeting of CDCA3.
Cell journal (2023-08-29)
Yongqiang Lai, Junhao Liu, Xiao Hu, Xiancheng Zeng, Peng Gao
ZUSAMMENFASSUNG
Owing to the lethality of liver cancer, it is considered as one of the devastating types of cancers across the globe. Consistently, the study was designed to elucidate the role and to explore the therapeutic implications of miR-145 in human liver cancer. In the current experimental study, gene expression was determined by RT-PCR analysis. Transfection of cancer cells was carried out using Lipofectamine 2000. The cell proliferation of liver cancer cells was estimated by MTT assay. Clonogenic assay was performed for analysis of colony forming potential of cancer cells. Flow cytometry was done to analyze the cell cycle phase distribution of cancer cells. Transwell chamber assay was performed to assess the motility of cancer cells. Western blotting was done to estimate the expression levels of proteins. Dual luciferase assay was performed for interaction analysis of miR-145 with CDCA3. The miR-145 expression was found to be downregulated in liver cancer cells. The transfection mediated overexpression of miR-145 inhibited the cancer cell proliferation and when miR-145 inhibitor was transfected, cancer cells showed higher proliferation rates. Enrichment of miR-145 levels led to cell cycle arrest at G2/M phase by inhibiting cyclin B1. miR-145 also restricted the migration and invasion of cancer cells. CDCA3 was shown to be the intracellular target of miR-145 and it was found that the inhibitory effects of miR-145 were modulated through CDCA3, intracellularly. The current study clearly revealed that there is a need to investigate the regulatory role of different molecular entities like microRNAs in cancer development to better understand mechanics behind this pathogenesis and design more effective combating strategies against cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-CDCA3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution