Merck
  • A Mesp1-dependent developmental breakpoint in transcriptional and epigenomic specification of early cardiac precursors.
A Mesp1-dependent developmental breakpoint in transcriptional and epigenomic specification of early cardiac precursors.
Development (Cambridge, England) (2023-03-31)
Alexis Leigh Krup, Sarah A B Winchester, Sanjeev S Ranade, Ayushi Agrawal, W Patrick Devine, Tanvi Sinha, Krishna Choudhary, Martin H Dominguez, Reuben Thomas, Brian L Black, Deepak Srivastava, Benoit G Bruneau
ZUSAMMENFASSUNG
Transcriptional networks governing cardiac precursor cell (CPC) specification are incompletely understood owing, in part, to limitations in distinguishing CPCs from non-cardiac mesoderm in early gastrulation. We leveraged detection of early cardiac lineage transgenes within a granular single-cell transcriptomic time course of mouse embryos to identify emerging CPCs and describe their transcriptional profiles. Mesp1, a transiently expressed mesodermal transcription factor, is canonically described as an early regulator of cardiac specification. However, we observed perdurance of CPC transgene-expressing cells in Mesp1 mutants, albeit mislocalized, prompting us to investigate the scope of the role of Mesp1 in CPC emergence and differentiation. Mesp1 mutant CPCs failed to robustly activate markers of cardiomyocyte maturity and crucial cardiac transcription factors, yet they exhibited transcriptional profiles resembling cardiac mesoderm progressing towards cardiomyocyte fates. Single-cell chromatin accessibility analysis defined a Mesp1-dependent developmental breakpoint in cardiac lineage progression at a shift from mesendoderm transcriptional networks to those necessary for cardiac patterning and morphogenesis. These results reveal Mesp1-independent aspects of early CPC specification and underscore a Mesp1-dependent regulatory landscape required for progression through cardiogenesis.