Merck
Sonic hedgehog is not a limb morphogen but acts as a trigger to specify all digits in mice.
Developmental cell (2022-08-18)
Jianjian Zhu, Rashmi Patel, Anna Trofka, Brian D Harfe, Susan Mackem
ZUSAMMENFASSUNG
Limb patterning by Sonic hedgehog (Shh), via either graded spatial or temporal signal integration, is a paradigm for "morphogen" function, yet how Shh instructs distinct digit identities remains controversial. Here, we bypass the Shh requirement in cell survival during outgrowth and demonstrate that a transient, early Shh pulse is both necessary and sufficient for normal mouse limb development. Shh response is only short range and is limited to the Shh-expressing zone during this time window. Shh patterns digits 1-3, anterior to this zone, by an indirect mechanism rather than direct spatial or temporal signal integration. Using a genetic relay-signaling assay, we discover that Shh also specifies digit 1/thumb (thought to be exclusively Shh independent) indirectly, and this finding implicates Shh in a unique regulatory hierarchy for digit 1 evolutionary adaptations such as opposable thumbs. This study illuminates Shh as a trigger for an indirect downstream network that becomes rapidly self-sustaining, with mechanistic relevance for limb development, regeneration, and evolution.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tamoxifen, ≥99%
Roche
cOmplete Mini Proteasehemmer-Cocktail, Tablets provided in a glass vial
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Roche
Anti-Digoxigenin-AP, Fab-Fragmente, from sheep
Sigma-Aldrich
Anti-Vinculin-Antikörper, Maus monoklonal, clone hVIN-1, purified from hybridoma cell culture