Merck
Alterations induced by the PML-RARα oncogene revealed by image cross correlation spectroscopy.
Biophysical journal (2022-10-06)
Elena Cerutti, Morgana D'Amico, Isotta Cainero, Pier Giuseppe Pelicci, Mario Faretta, Gaetano Ivan Dellino, Alberto Diaspro, Luca Lanzanò
ZUSAMMENFASSUNG
The molecular mechanisms that underlie oncogene-induced genomic damage are still poorly understood. To understand how oncogenes affect chromatin architecture, it is important to visualize fundamental processes such as DNA replication and transcription in intact nuclei and quantify the alterations of their spatiotemporal organization induced by oncogenes. Here, we apply superresolution microscopy in combination with image cross correlation spectroscopy to the U937-PR9 cell line, an in vitro model of acute promyelocytic leukemia that allows us to activate the expression of the PML-RARα oncogene and analyze its effects on the spatiotemporal organization of functional nuclear processes. More specifically, we perform Tau-stimulated emission depletion imaging, a superresolution technique based on the concept of separation of photons by lifetime tuning. Tau-stimulated emission depletion imaging is combined with a robust image analysis protocol that quickly produces a value of colocalization fraction on several hundreds of single cells and allows observation of cell-to-cell variability. Upon activation of the oncogene, we detect a significant increase in the fraction of transcription sites colocalized with PML/PML-RARα. This increase of colocalization can be ascribed to oncogene-induced disruption of physiological PML bodies and the abnormal occurrence of a relatively large number of PML-RARα microspeckles. We also detect a significant cell-to-cell variability of this increase of colocalization, which can be ascribed, at least in part, to a heterogeneous response of the cells to the activation of the oncogene. These results prove that our method efficiently reveals oncogene-induced alterations in the spatial organization of nuclear processes and suggest that the abnormal localization of PML-RARα could interfere with the transcription machinery, potentially leading to DNA damage and genomic instability.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Kaninchen-IgG - Atto 647N, 1 mg/mL IgG
Sigma-Aldrich
Anti-Maus-IgG - Atto 647N, contains 50% glycerol as stabilizer
Sigma-Aldrich
Anti-Maus-IgG – Atto 594 in Ziege hergestellte Antikörper, ~1 mg/mL protein, affinity isolated antibody
Sigma-Aldrich
Anti-Kaninchen-IgG – Atto 594 in Ziege hergestellte Antikörper