Merck
  • Derivatives of 17-(2-methylallyl)-substituted noroxymorphone: variation of the delta address and its effects on affinity and selectivity for the delta opioid receptor.
Derivatives of 17-(2-methylallyl)-substituted noroxymorphone: variation of the delta address and its effects on affinity and selectivity for the delta opioid receptor.
Bioorganic & medicinal chemistry letters (2001-10-13)
T Ullrich, C M Dersch, R B Rothman, A E Jacobson, K C Rice
ZUSAMMENFASSUNG
In an effort to establish the importance of the N-(2-methylallyl) substituent in the noroxymorphone series, several derivatives have been synthesized, retaining that N-substituent and modifying the delta address moiety. A few compounds showed moderate binding affinity and selectivity for the delta receptor; none displayed a pharmacological profile as exceptional as N-(2-methylallyl)noroxymorphindole. A second study showed that 3-O-methylation of all derivatives decreases binding affinity. The present results indicate that only a combination of the N-(2-methylallyl) group and an indole delta address provided high selectivity for the delta receptor.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
SDM25N, ≥98% (HPLC)