Merck
  • Escape Mutations in NS4B Render Dengue Virus Insensitive to the Antiviral Activity of the Paracetamol Metabolite AM404.
Escape Mutations in NS4B Render Dengue Virus Insensitive to the Antiviral Activity of the Paracetamol Metabolite AM404.
Antimicrobial agents and chemotherapy (2016-02-10)
Koen W R van Cleef, Gijs J Overheul, Michael C Thomassen, Jenni M Marjakangas, Ronald P van Rij
ZUSAMMENFASSUNG
Despite the enormous disease burden associated with dengue virus infections, a licensed antiviral drug is lacking. Here, we show that the paracetamol (acetaminophen) metabolite AM404 inhibits dengue virus replication. Moreover, we find that mutations in NS4B that were previously found to confer resistance to the antiviral compounds NITD-618 and SDM25N also render dengue virus insensitive to AM404. Our work provides further support for NS4B as a direct or indirect target for antiviral drug development.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
SDM25N, ≥98% (HPLC)