Merck
  • Cardiac epigenetic changes in VEGF signaling genes associate with myocardial microvascular rarefaction in experimental chronic kidney disease.
Cardiac epigenetic changes in VEGF signaling genes associate with myocardial microvascular rarefaction in experimental chronic kidney disease.
American journal of physiology. Heart and circulatory physiology (2022-11-12)
Alfonso Eirin, Alejandro R Chade
ZUSAMMENFASSUNG
Chronic kidney disease (CKD) is common in patients with heart failure and often results in left ventricular diastolic dysfunction (LVDD). However, the mechanisms responsible for cardiac damage in CKD-LVDD remain to be elucidated. Epigenetic alterations may impose long-lasting effects on cellular transcription and function, but their exact role in CKD-LVDD is unknown. We investigate whether changes in cardiac site-specific DNA methylation profiles might be implicated in cardiac abnormalities in CKD-LVDD. CKD-LVDD and normal control pigs (n = 6 each) were studied for 14 wk. Renal and cardiac hemodynamics were quantified by multidetector CT and echocardiography. In randomly selected pigs (n = 3/group), cardiac site-specific 5-methylcytosine (5mC) immunoprecipitation (MeDIP)- and mRNA-sequencing (seq) were performed, followed by integrated (MeDiP-seq/mRNA-seq analysis), and confirmatory ex vivo studies. MeDIP-seq analysis revealed 261 genes with higher (fold change > 1.4; P < 0.05) and 162 genes with lower (fold change < 0.7; P < 0.05) 5mC levels in CKD-LVDD versus normal pigs, which were primarily implicated in vascular endothelial growth factor (VEGF)-related signaling and angiogenesis. Integrated MeDiP-seq/mRNA-seq analysis identified a select group of VEGF-related genes in which 5mC levels were higher, but mRNA expression was lower in CKD-LVDD versus normal pigs. Cardiac VEGF signaling gene and VEGF protein expression were blunted in CKD-LVDD compared with controls and were associated with decreased subendocardial microvascular density. Cardiac epigenetic changes in VEGF-related genes are associated with impaired angiogenesis and cardiac microvascular rarefaction in swine CKD-LVDD. These observations may assist in developing novel therapies to ameliorate cardiac damage in CKD-LVDD.NEW & NOTEWORTHY Chronic kidney disease (CKD) often leads to left ventricular diastolic dysfunction (LVDD) and heart failure. Using a novel translational swine model of CKD-LVDD, we characterize the cardiac epigenetic landscape, identifying site-specific 5-methylcytosine changes in vascular endothelial growth factor (VEGF)-related genes associated with impaired angiogenesis and cardiac microvascular rarefaction. These observations shed light on the mechanisms of cardiac microvascular damage in CKD-LVDD and may assist in developing novel therapies for these patients.