Merck
  • CD70 is a therapeutic target upregulated in EMT-associated EGFR tyrosine kinase inhibitor resistance.
CD70 is a therapeutic target upregulated in EMT-associated EGFR tyrosine kinase inhibitor resistance.
Cancer cell (2023-02-15)
Monique B Nilsson, Yan Yang, Simon Heeke, Sonia A Patel, Alissa Poteete, Hibiki Udagawa, Yasir Y Elamin, Cesar A Moran, Yukie Kashima, Thiruvengadam Arumugam, Xiaoxing Yu, Xiaoyang Ren, Lixia Diao, Li Shen, Qi Wang, Minying Zhang, Jacqulyne P Robichaux, Chunhua Shi, Allyson N Pfeil, Hai Tran, Don L Gibbons, Jason Bock, Jing Wang, John D Minna, Susumu S Kobayashi, Xiuning Le, John V Heymach
ZUSAMMENFASSUNG
Effective therapeutic strategies are needed for non-small cell lung cancer (NSCLC) patients with epidermal growth factor receptor (EGFR) mutations that acquire resistance to EGFR tyrosine kinase inhibitors (TKIs) mediated by epithelial-to-mesenchymal transition (EMT). We investigate cell surface proteins that could be targeted by antibody-based or adoptive cell therapy approaches and identify CD70 as being highly upregulated in EMT-associated resistance. Moreover, CD70 upregulation is an early event in the evolution of resistance and occurs in drug-tolerant persister cells (DTPCs). CD70 promotes cell survival and invasiveness, and stimulation of CD70 triggers signal transduction pathways known to be re-activated with acquired TKI resistance. Anti-CD70 antibody drug conjugates (ADCs) and CD70-targeting chimeric antigen receptor (CAR) T cell and CAR NK cells show potent activity against EGFR TKI-resistant cells and DTPCs. These results identify CD70 as a therapeutic target for EGFR mutant tumors with acquired EGFR TKI resistance that merits clinical investigation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-15, ascites fluid
Sigma-Aldrich
Monoklonaler Anti-Vinculin-Antikörper in Maus hergestellte Antikörper, clone hVIN-1, ascites fluid