Merck
  • Hypoxia inducible factor-1? regulates microglial innate immune memory and the pathology of Parkinson?s disease
Hypoxia inducible factor-1? regulates microglial innate immune memory and the pathology of Parkinson?s disease
Journal of Neuroinflammation (2024)
Dong, et al.