Merck
  • Comparison of vascular endothelial growth factor and fibroblast growth factor-2 in a swine model of endothelial dysfunction.
Comparison of vascular endothelial growth factor and fibroblast growth factor-2 in a swine model of endothelial dysfunction.
European journal of cardio-thoracic surgery : official journal of the European Association for Cardio-thoracic Surgery (2008-01-19)
Munir Boodhwani, Pierre Voisine, Marc Ruel, Neel R Sodha, Jun Feng, Shu-Hua Xu, Cesario Bianchi, Frank W Sellke
ZUSAMMENFASSUNG
Growth factor based angiogenesis, with or without cell therapy, is a promising therapeutic modality for patients with coronary artery disease. We compared the relative efficacies of surgically delivered vascular endothelial growth factor (VEGF) and fibroblast growth factor-2 (FGF-2) in a swine model of hypercholesterolemia-induced endothelial dysfunction which captures many of the pathophysiologic abnormalities of human coronary disease. Yucatan mini-swine (20-30 kg), fed a high cholesterol diet (total 20 weeks), underwent circumflex ameroid placement to create chronic myocardial ischemia, followed three weeks later by perivascular administration of VEGF (2 microg; n=6), FGF-2 (100 microg; n=6), or placebo (n=7) in the ischemic territory. Normocholesterolemic animals (n=7) served as controls. Four weeks later, endothelial function, collateral-dependent perfusion, as well as myocardial protein and mRNA levels of angiogenic mediators were assessed. Endothelial dysfunction was observed in all hypercholesterolemic animals as impaired microvessel relaxation in response to adenosine diphosphate and VEGF. VEGF administration improved baseline-adjusted collateral-dependent perfusion at rest (-0.03+/-0.05 vs -0.12+/-0.04, VEGF vs placebo, p=0.09), but FGF-2 delivery caused a significantly greater improvement in perfusion compared to either group (+0.15+/-0.03, p<0.05 vs HC-placebo and HC-VEGF) at rest. Molecular analysis revealed increased eNOS expression (135%+/-8%, p=0.03 vs placebo) in all growth factor treated animals and increased expression of FGF-2 receptor, FGFR1 (65+/-26%, p=0.04 vs placebo), in FGF-2 treated animals. No significant changes were demonstrated in other angiogenic mediators including Akt, Syndecan-4. In the setting of hypercholesterolemic endothelial dysfunction, FGF-2 is more effective than VEGF at enhancing collateral-dependent perfusion and thus, may be a better candidate than VEGF for angiogenic therapy in patients with end-stage CAD.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
TRI-Reagenz®, For processing tissues, cells cultured in monolayer or cell pellets
Sigma-Aldrich
TRI-Reagenz®
Sigma-Aldrich
TRI-Reagenz®, LS, For processing fluid samples such as cell suspensions, CSF, and amniotic fluid.
Sigma-Aldrich
TRI-Reagenz®, BD, For processing whole blood, plasma, or serum.