Merck
  • Sex differences in the effect of the FKBP5 inhibitor SAFit2 on anxiety and stress-induced reinstatement following cocaine self-administration.
Sex differences in the effect of the FKBP5 inhibitor SAFit2 on anxiety and stress-induced reinstatement following cocaine self-administration.
Neurobiology of stress (2020-12-22)
Krista L Connelly, Cassandra C Wolsh, Jeffrey L Barr, Michael Bauder, Felix Hausch, Ellen M Unterwald
ZUSAMMENFASSUNG
Cocaine use and withdrawal prompt stress system responses. Stress and the negative affective state produced by cocaine withdrawal are major triggers for relapse. FKBP5 is a co-chaperone of the glucocorticoid receptor and regulates HPA axis negative feedback. The role of FKBP5 in cocaine-related behaviors has not been studied. The FKBP5 inhibitor SAFit2 was used to examine the role of FKBP5 in anxiety-like behavior during early cocaine withdrawal and in stress-induced reinstatement following cocaine self-administration in male and female rats. Withdrawal from cocaine self-administration resulted in heightened anxiety-like behavior in female rats, which was significantly attenuated by SAFit2 administration. SAFit2 pretreatment prior to stress-induced reinstatement to cocaine seeking significantly reduced active lever presses of males. In female rats, SAFit2 administration prevented stress-induced reinstatement for rats in metestrus or diestrus, but not proestrus or estrus phases at the time of reinstatement. These data suggest an important role for FKBP5 in stress-related behaviors following cocaine self-administration, particularly in females.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
SAFit2, ≥98% (HPLC)