Merck
  • The selective FKBP51 inhibitor SAFit2 reduces alcohol consumption and reinstatement of conditioned alcohol effects in mice.
The selective FKBP51 inhibitor SAFit2 reduces alcohol consumption and reinstatement of conditioned alcohol effects in mice.
Addiction biology (2019-06-08)
Loretta König, Liubov S Kalinichenko, Sabine E Huber, Andreas M Voll, Michael Bauder, Johannes Kornhuber, Felix Hausch, Christian P Müller
ZUSAMMENFASSUNG
There is still no widely effective pharmacotherapy for alcohol addiction available in the clinic. FK506-binding protein 51 (FKBP51) is a negative regulator of the glucocorticoid receptor signaling pathway that regulates the stress-induced glucocorticoid feedback circuit. Here we asked whether selective inhibitors of FKBP51, exemplified by SAFit2, may serve as a new pharmacological strategy to reduce alcohol consumption and conditioned alcohol effects in a mouse model. We report that a relatively short treatment with SAFit2 (20 mg/kg, ip) reduces ongoing 16 vol% alcohol consumption when administered during free access to alcohol in a two-bottle free-choice test. SAFit2 was also able to reduce alcohol consumption when given during an abstinence period immediately before relapse. In contrast, SAFit2 did not affect alcohol consumption when given during a relapse period after repeated withdrawal from alcohol. SAFit2 (10 and 20 mg/kg, ip) showed no effects when used in an intermittent drinking schedule. When 20 vol% alcohol was only available every other day, SAFit2 had no effect on drinking, no matter whether given during a drinking episode or the day before. SAFit2 (2 and 20 mg/kg, ip) did not affect the expression of an alcohol-induced conditioned place preference (CPP). However, SAFit2 was able to inhibit alcohol-induced reinstatement of an extinguished CPP in a dose-dependent way. Altogether, these data may suggest pharmacological inhibition of FKBP51 as a viable strategy to reduce alcohol seeking and consumption.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
SAFit2, ≥98% (HPLC)