Merck
KDM6A epigenetically regulates subtype plasticity in small cell lung cancer.
Nature cell biology (2023-08-18)
Leslie Duplaquet, Yixiang Li, Matthew A Booker, Yingtian Xie, Sarah Naomi Olsen, Radhika A Patel, Deli Hong, Charlie Hatton, Thomas Denize, Emily Walton, Yasmin N Laimon, Rong Li, Yijia Jiang, Roderick T Bronson, Jackson Southard, Shuqiang Li, Sabina Signoretti, Xintao Qiu, Paloma Cejas, Scott A Armstrong, Henry W Long, Michael Y Tolstorukov, Michael C Haffner, Matthew G Oser
ZUSAMMENFASSUNG
Small cell lung cancer (SCLC) exists broadly in four molecular subtypes: ASCL1, NEUROD1, POU2F3 and Inflammatory. Initially, SCLC subtypes were thought to be mutually exclusive, but recent evidence shows intra-tumoural subtype heterogeneity and plasticity between subtypes. Here, using a CRISPR-based autochthonous SCLC genetically engineered mouse model to study the consequences of KDM6A/UTX inactivation, we show that KDM6A inactivation induced plasticity from ASCL1 to NEUROD1 resulting in SCLC tumours that express both ASCL1 and NEUROD1. Mechanistically, KDM6A normally maintains an active chromatin state that favours the ASCL1 subtype with its loss decreasing H3K4me1 and increasing H3K27me3 at enhancers of neuroendocrine genes leading to a cell state that is primed for ASCL1-to-NEUROD1 subtype switching. This work identifies KDM6A as an epigenetic regulator that controls ASCL1 to NEUROD1 subtype plasticity and provides an autochthonous SCLC genetically engineered mouse model to model ASCL1 and NEUROD1 subtype heterogeneity and plasticity, which is found in 35-40% of human SCLCs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Roche
cOmplete Mini Proteasehemmer-Cocktail, Tablets provided in a glass vial
Sigma-Aldrich
Monoklonaler Anti-Vinculin-Antikörper in Maus hergestellte Antikörper, clone hVIN-1, ascites fluid
Sigma-Aldrich
Hydrocortison, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Monoklonaler Anti-β-Aktin−Peroxidase-Antikörper der Maus in Maus hergestellte Antikörper, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
PF-9363, ≥98% (HPLC)
Sigma-Aldrich
PF-9363, ≥98% (HPLC)