Merck
Acetyl-CoA biosynthesis drives resistance to histone acetyltransferase inhibition.
Nature chemical biology (2023-05-02)
Timothy R Bishop, Chitra Subramanian, Eric M Bilotta, Leopold Garnar-Wortzel, Anissa R Ramos, Yuxiang Zhang, Joshua N Asiaban, Christopher J Ott, Charles O Rock, Michael A Erb
ZUSAMMENFASSUNG
Histone acetyltransferases (HATs) are implicated as both oncogene and nononcogene dependencies in diverse human cancers. Acetyl-CoA-competitive HAT inhibitors have emerged as potential cancer therapeutics and the first clinical trial for this class of drugs is ongoing (NCT04606446). Despite these developments, the potential mechanisms of therapeutic response and evolved drug resistance remain poorly understood. Having discovered that multiple regulators of de novo coenzyme A (CoA) biosynthesis can modulate sensitivity to CBP/p300 HAT inhibition (PANK3, PANK4 and SLC5A6), we determined that elevated acetyl-CoA concentrations can outcompete drug-target engagement to elicit acquired drug resistance. This not only affects structurally diverse CBP/p300 HAT inhibitors, but also agents related to an investigational KAT6A/B HAT inhibitor that is currently in Phase 1 clinical trials. Altogether, this work uncovers CoA metabolism as an unexpected liability of anticancer HAT inhibitors and will therefore buoy future efforts to optimize the efficacy of this new form of targeted therapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Acetyl-Histone-H3(Lys23)-Antikörper, serum, Upstate®
Sigma-Aldrich
Anti-Acetyl-Histon-H3(Lys18)-Antikörper, serum, Upstate®
Sigma-Aldrich
PF-9363, ≥98% (HPLC)
Sigma-Aldrich
PF-9363, ≥98% (HPLC)