Merck
  • Promotion of Knee Cartilage Degradation by I?B Kinase ? in the Pathogenesis of Osteoarthritis in Human and Murine Models
Promotion of Knee Cartilage Degradation by I?B Kinase ? in the Pathogenesis of Osteoarthritis in Human and Murine Models
Arthritis & Rheumatology (Hoboken, N.J.) (2023)