Merck
Folic Acid and PEI Modified Mesoporous Silica for Targeted Delivery of Curcumin.
Pharmaceutics (2019-08-28)
Xiaoxiao Sun, Nan Wang, Li-Ye Yang, Xiao-Kun Ouyang, Fangfang Huang
ZUSAMMENFASSUNG
Nano anti-cancer drug carriers loaded with antineoplastic drugs can achieve targeted drug delivery, which enriches drugs at tumor sites and reduces the toxic side effects in normal tissues. Mesoporous silica nanoparticles (MSN) are good nano drug carriers, as they have large specific surface areas, adjustable pore sizes, easily modifiable surfaces, and good biocompatibility. In this work, polyethyleneimine (PEI) grafted MSN were modified with folic acid (FA) as an active target molecule using chemical methods. The product was characterized by SEM, TEM, Zetasizer nano, FTIR, and an N2 adsorption and desorption test. MSN-PEI-FA are porous nano particles with an average particle size of approximately 100 nm. In addition, the loading rate and release behavior of MSN-PEI-FA were studied with curcumin as a model drug. The results show that when loading curcumin to MSN-PEI-FA at 7 mg and 0.1 g, respectively, the encapsulation efficiency was 90% and the cumulative release rate reached more than 50% within 120 h at pH = 5. This drug delivery system is suitable for loading fat-soluble antineoplastic drugs for sustained release and pH sensitive delivery.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Folic acid modified polyethylenimine solution, low molecular weight, 10 mg/mL (aqueous solution), suitable for biomedical research
Sigma-Aldrich
Folic acid modified polyethylenimine solution, high molecular weight, 10 mg/ml aqueous solution, suitable for biomedical research