Merck
  • Effect of the size of zona pellucida opening by laser assisted hatching on clinical outcome of frozen cleaved embryos that were cultured to blastocyst after thawing in women with multiple implantation failures of embryo transfer: a retrospective study.
Effect of the size of zona pellucida opening by laser assisted hatching on clinical outcome of frozen cleaved embryos that were cultured to blastocyst after thawing in women with multiple implantation failures of embryo transfer: a retrospective study.
Journal of assisted reproduction and genetics (2008-03-28)
Kenichiro Hiraoka, Megumi Fuchiwaki, Kaori Hiraoka, Toshitaka Horiuchi, Tomoyo Murakami, Masayuki Kinutani, Kazuo Kinutani
ZUSAMMENFASSUNG
To evaluate the effect of the size of zona pellucida opening by laser assisted hatching for frozen cleaved embryo that were thawed after both fresh and frozen cleaved embryo transfer cycles failed and were cultured to blastocyst after thawing in patients with multiple implantation failures. Of 101 consecutive procedures (October 2003 to June 2006), 30 patients declined to perform assisted hatching and were selected as control group, 40 patients had 40 microm opening of the zona (October 2003 to January 2005), 31 patients had 50% of the zona opening (February 2005 to June 2006). The pregnancy, implantation and delivery rates were significantly higher in 50% opening group (74%, 52% and 65%) compared to control group (17%, 10% and 13%; P < 0.01) and 40 microm opening group (43%, 27% and 38%; P < 0.04). The size of the zona pellucida opening may affect the outcome of frozen cleaved embryo transfer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethylenglycol, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethylenglycol, anhydrous, 99.8%
Sigma-Aldrich
Ethylenglycol, spectrophotometric grade, ≥99%
Supelco
Ethylenglycol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethylenglycol, analytical standard
Sigma-Aldrich
Ethylenglycol, BioUltra, ≥99.5% (GC)