Merck
Structural basis for substrate promiscuity of dCK.
Journal of molecular biology (2008-04-02)
Elisabetta Sabini, Saugata Hazra, Stephan Ort, Manfred Konrad, Arnon Lavie
ZUSAMMENFASSUNG
Deoxycytidine kinase (dCK) is an essential nucleoside kinase critical for the production of nucleotide precursors for DNA synthesis. This enzyme catalyzes the initial conversion of the nucleosides deoxyadenosine (dA), deoxyguanosine (dG), and deoxycytidine (dC) into their monophosphate forms, with subsequent phosphorylation to the triphosphate forms performed by additional enzymes. Several nucleoside analog prodrugs are dependent on dCK for their pharmacological activation, and even nucleosides of the non-physiological L-chirality are phosphorylated by dCK. In addition to accepting dC and purine nucleosides (and their analogs) as phosphoryl acceptors, dCK can utilize either ATP or UTP as phosphoryl donors. To unravel the structural basis for substrate promiscuity of dCK at both the nucleoside acceptor and nucleotide donor sites, we solved the crystal structures of the enzyme as ternary complexes with the two enantiomeric forms of dA (D-dA, or L-dA), with either UDP or ADP bound to the donor site. The complexes with UDP revealed an open state of dCK in which the nucleoside, either D-dA or L-dA, is surprisingly bound in a manner not consistent with catalysis. In contrast, the complexes with ADP, with either D-dA or L-dA, adopted a closed and catalytically competent conformation. The differential states adopted by dCK in response to the nature of the nucleotide were also detected by tryptophan fluorescence experiments. Thus, we are in the unique position to observe differential effects at the acceptor site due to the nature of the nucleotide at the donor site, allowing us to rationalize the different kinetic properties observed with UTP to those with ATP.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Mineralöl, BioReagent, Molecular Biology, light oil
Sigma-Aldrich
Mineralöl, light oil, (neat)
Sigma-Aldrich
Mineralöl, light oil, suitable for mouse embryo cell culture, BioXtra
Sigma-Aldrich
Mineralöl, light
Sigma-Aldrich
Mineralöl, suitable for preparation of Nujol mulls for infrared spectroscopy, light oil
Sigma-Aldrich
Mineralöl, heavy
Sigma-Aldrich
Mineralöl, meets USP testing specifications
Sigma-Aldrich
Mineralöl, PCR Reagent
Sigma-Aldrich
Mineralöl, BioUltra, Molecular Biology