Merck
Early molecular layer interneuron hyperactivity triggers Purkinje neuron degeneration in SCA1.
Neuron (2023-06-16)
Federica Pilotto, Christopher Douthwaite, Rim Diab, XiaoQian Ye, Zahraa Al Qassab, Christoph Tietje, Meriem Mounassir, Adolfo Odriozola, Aishwarya Thapa, Ronald A M Buijsen, Sophie Lagache, Anne-Christine Uldry, Manfred Heller, Stefan Müller, Willeke M C van Roon-Mom, Benoît Zuber, Sabine Liebscher, Smita Saxena
ZUSAMMENFASSUNG
Toxic proteinaceous deposits and alterations in excitability and activity levels characterize vulnerable neuronal populations in neurodegenerative diseases. Using in vivo two-photon imaging in behaving spinocerebellar ataxia type 1 (Sca1) mice, wherein Purkinje neurons (PNs) degenerate, we identify an inhibitory circuit element (molecular layer interneurons [MLINs]) that becomes prematurely hyperexcitable, compromising sensorimotor signals in the cerebellum at early stages. Mutant MLINs express abnormally elevated parvalbumin, harbor high excitatory-to-inhibitory synaptic density, and display more numerous synaptic connections on PNs, indicating an excitation/inhibition imbalance. Chemogenetic inhibition of hyperexcitable MLINs normalizes parvalbumin expression and restores calcium signaling in Sca1 PNs. Chronic inhibition of mutant MLINs delayed PN degeneration, reduced pathology, and ameliorated motor deficits in Sca1 mice. Conserved proteomic signature of Sca1 MLINs, shared with human SCA1 interneurons, involved the higher expression of FRRS1L, implicated in AMPA receptor trafficking. We thus propose that circuit-level deficits upstream of PNs are one of the main disease triggers in SCA1.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-GABA in Kaninchen hergestellte Antikörper, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Glutamatrezeptor-2-Antikörper, extrazellulär, Klon 6C4, clone 6C4, Chemicon®, from mouse
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-MAP2-Antikörper, Chemicon®, from chicken