Merck
DNA damage induces nuclear envelope rupture through ATR-mediated phosphorylation of lamin A/C.
Molecular cell (2023-10-14)
Marton Tibor Kovacs, Marie Vallette, Pauline Wiertsema, Florent Dingli, Damarys Loew, Guilherme Pedreira de Freitas Nader, Matthieu Piel, Raphael Ceccaldi
ZUSAMMENFASSUNG
The integrity of the nuclear envelope (NE) is essential for maintaining the structural stability of the nucleus. Rupture of the NE has been frequently observed in cancer cells, especially in the context of mechanical challenges, such as physical confinement and migration. However, spontaneous NE rupture events, without any obvious physical challenges to the cell, have also been described. The molecular mechanism(s) of these spontaneous NE rupture events remain to be explored. Here, we show that DNA damage and subsequent ATR activation leads to NE rupture. Upon DNA damage, lamin A/C is phosphorylated in an ATR-dependent manner, leading to changes in lamina assembly and, ultimately, NE rupture. In addition, we show that cancer cells with intrinsic DNA repair defects undergo frequent events of DNA-damage-induced NE rupture, which renders them extremely sensitive to further NE perturbations. Exploiting this NE vulnerability could provide a new angle to complement traditional, DNA-damage-based chemotherapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-phospho-Histon H2A.X (Ser139)-Antikörper, Klon JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-74, ascites fluid
Sigma-Aldrich
Etoposid, A cell-permeable derivative of podophyllotoxin that acts as a topoisomerase II inhibitor (IC50 = 59.2 µM) has major activity against a number of tumors, including germ cell neoplasms, small cell lung cancer, and malignant lymphoma.
Sigma-Aldrich
Trichostatin A, Ready Made Solution, 5 mM in DMSO, from Streptomyces sp.
Sigma-Aldrich
CK-666, ≥98% (HPLC), powder
Sigma-Aldrich
Camptothecin, Camptotheca acuminata, A cell-permeable DNA topoisomerase I inhibitor.
Sigma-Aldrich
Hydrocortison, Steroid hormone of the adrenal cortex with anti-inflammatory properties.
Sigma-Aldrich
Anti-IL-2 Rα Antibody, clone 7G7/B6, clone 7G7/B6, Upstate®, from mouse