Merck
Forkhead box protein D2 suppresses colorectal cancer by reprogramming enhancer interactions.
Nucleic acids research (2023-05-09)
Hyo-Min Kim, Byunghee Kang, Sohyun Park, Hyorim Park, Chan Johng Kim, Hyeonji Lee, Mijoung Yoo, Mi-Na Kweon, Sin-Hyeog Im, Tae Il Kim, Tae-Young Roh
ZUSAMMENFASSUNG
Somatic stem cells contribute to normal tissue homeostasis, and their epigenomic features play an important role in regulating tissue identities or developing disease states. Enhancers are one of the key players controlling chromatin context-specific gene expression in a spatial and temporal manner while maintaining tissue homeostasis, and their dysregulation leads to tumorigenesis. Here, epigenomic and transcriptomic analyses reveal that forkhead box protein D2 (FOXD2) is a hub for the gene regulatory network exclusive to large intestinal stem cells, and its overexpression plays a significant role in colon cancer regression. FOXD2 is positioned at the closed chromatin and facilitates mixed-lineage leukemia protein-4 (MLL4/KMT2D) binding to deposit H3K4 monomethylation. De novo FOXD2-mediated chromatin interactions rewire the regulation of p53-responsive genes and induction of apoptosis. Taken together, our findings illustrate the novel mechanistic details of FOXD2 in suppressing colorectal cancer growth and suggest its function as a chromatin-tuning factor and a potential therapeutic target for colorectal cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Y-27632 -dihydrochlorid, ≥98% (HPLC)
Roche
cOmplete-ULTRA-Mini-Tabletten, Protease-Inhibitor-Cocktail in EASYpacks , Tablets supplied in foil blister packs.
Sigma-Aldrich
ANTI-FLAG® antibody, Rat monoclonal, clone 6F7, purified from hybridoma cell culture
Sigma-Aldrich
Anti-FOXD2 antibody produced in rabbit, IgG fraction of antiserum