Merck
  • Expression mapping of GREM1 and functional contribution of its secreting cells in the brain using transgenic mouse models.
Expression mapping of GREM1 and functional contribution of its secreting cells in the brain using transgenic mouse models.
Experimental neurology (2023-12-11)
Peijia Yao, Xueli Liu, Qiang Miao, Changxue Li, Huaixiang Zhou, Huiliang Li, Xinliang Mao, Xiaoyi Fang, Ningning Li
ZUSAMMENFASSUNG
GREMLIN1 (GREM1) is a secreted protein that antagonizes bone morphogenetic proteins (BMPs). While abnormal GREM1 expression has been reported to cause behavioral defects in postpartum mice, the spatial and cellular distribution of GREM1 in the brain and the influence of the GREM1-secreting cells on brain function and behavior remain unclear. To address this, we designed a genetic cassette incorporating a 3×Flag-TeV-HA-T2A-tdTomato sequence, resulting in the creation of a novel Grem1Tag mouse model, expressing an epitope tag (3×Flag-TeV-HA-T2A) followed by a fluorescent reporter (tdTomato) under the control of the endogenous Grem1 promoter. This design facilitated precise tracking of the cell origin and distribution of GREM1 in the brain using tdTomato and Flag (or HA) markers, respectively. We confirmed that the Grem1Tag mouse exhibited normal motor, cognitive, and social behaviors at postnatal 60 days (P60), compared with C57BL/6J controls. Through immunofluorescence staining, we comprehensively mapped the distribution of GREM1-secreting cells across the central nervous system. Pervasive GREM1 expression was observed in the cerebral cortex (Cx), medulla, pons, and cerebellum, with the highest levels in the Cx region. Notably, within the Cx, GREM1 was predominantly secreted by excitatory neurons, particularly those expressing calcium/calmodulin-dependent protein kinase II alpha (Camk2a), while inhibitory neurons (parvalbumin-positive, PV+) and glial cells (oligodendrocytes, astrocytes, and microglia) showed little or no GREM1 expression. To delineate the functional significance of GREM1-secreting cells, a selective ablation at P42 using a diphtheria toxin A (DTA) system resulted in increased anxiety-like behavior and impaired memory in mice. Altogether, our study harnessing the Grem1Tag mouse model reveals the spatial and cellular localization of GREM1 in the mouse brain, shedding light on the involvement of GREM1-secreting cells in modulating brain function and behavior. Our Grem1Tag mouse serves as a valuable tool for further exploring the precise role of GREM1 in brain development and disease.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Myelin-Oligodendrozyten-Glykoprotein (MOG)-Antikörper, clone 8-18C5, Chemicon®, from mouse