Merck
LKB1 signaling is altered in skeletal muscle of a Duchenne muscular dystrophy mouse model.
Disease models & mechanisms (2023-07-10)
Brigida Boccanegra, Paola Mantuano, Elena Conte, Alessandro Giovanni Cerchiara, Lisamaura Tulimiero, Raffaella Quarta, Erika Caputo, Francesca Sanarica, Monica Forino, Valeria Spadotto, Ornella Cappellari, Gianluca Fossati, Christian Steinkühler, Annamaria De Luca
ZUSAMMENFASSUNG
The potential role of liver kinase B1 (LKB1) in the altered activation of the master metabolic and epigenetic regulator adenosine monophosphate-activated protein kinase (AMPK) in Duchenne muscular dystrophy has not been investigated so far. Hence, we analyzed both gene and protein levels of LKB1 and its related targets in gastrocnemius muscles of adult C57BL/10 mdx mice and D2 mdx mice, a model with a more severe dystrophic phenotype, as well as the sensitivity of the LKB1-AMPK pathway to AMPK activators, such as chronic exercise. Our data show, for the first time, a reduction in the levels of LKB1 and accessory proteins, MO25 and STRADα, in both mdx strains versus the respective wild type, which was further impaired by exercise, in parallel with a lack of further phosphorylation of AMPK. The AMPK-like kinase salt-inducible kinase (SIK) and class II histone deacetylases, along with expression of the HDAC target gene Mef2c, were also altered, supporting an impairment of LKB1-SIK-class II histone deacetylase signaling. Our results demonstrate that LKB1 may be involved in dystrophic progression, paving the way for future preclinical studies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler Anti-Vinculin-Antikörper in Maus hergestellte Antikörper, clone hVIN-1, ascites fluid
Sigma-Aldrich
Anti-Maus-IgG (Gesamtmolekül)-Peroxidase in Kaninchen hergestellte Antikörper, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-LYK5 antibody produced in rabbit, affinity isolated antibody