Merck
  • Eicosapentaenoic acid increases proportion of type 1 muscle fibers through PPARδ and AMPK pathways in rats.
Eicosapentaenoic acid increases proportion of type 1 muscle fibers through PPARδ and AMPK pathways in rats.
iScience (2024-05-23)
Yusuke Komiya, Yuka Sakazaki, Tsuyoshi Goto, Fuminori Kawabata, Takahiro Suzuki, Yusuke Sato, Shoko Sawano, Mako Nakamura, Ryuichi Tatsumi, Yoshihide Ikeuchi, Keizo Arihara, Wataru Mizunoya
ZUSAMMENFASSUNG
Muscle fiber type composition (% slow-twitch and % fast-twitch fibers) is associated with metabolism, with increased slow-twitch fibers alleviating metabolic disorders. Previously, we reported that dietary fish oil intake induced a muscle fiber-type transition in a slower direction in rats. The aim of this study was to determine the functionality of eicosapentaenoic acid (EPA), a unique fatty acid in fish oil, to skeletal muscle fiber type and metabolism in rats. Here, we showed that dietary EPA promotes whole-body oxidative metabolism and improves muscle function by increasing proportion of slow-twitch type 1 fibers in rats. Transcriptomic and metabolomic analyses revealed that EPA supplementation activated the peroxisome proliferator-activated receptor δ (PPARδ) and AMP-activated protein kinase (AMPK) pathways in L6 myotube cultures, which potentially increasing slow-twitch fiber share. This highlights the role of EPA as an exercise-mimetic dietary component that improves metabolism and muscle function, with potential benefits for health and athletic performance.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Rosiglitazon
Sigma-Aldrich
Linolsäure, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Monoklonaler Anti-Myosin-Antikörper (skelettal, Fast) in Maus hergestellte Antikörper, clone MY-32, ascites fluid
Sigma-Aldrich
Anti-Aktin-Antikörper, Klon C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
Monoklonaler Anti-Myosin-Antikörper (skelettal, Slow) in Maus hergestellte Antikörper, clone NOQ7.5.4D, ascites fluid
Sigma-Aldrich
WY-14643