Merck
L-type Ca2+ channel function is linked to dystrophin expression in mammalian muscle.
PloS one (2008-06-03)
Oliver Friedrich, Frederic von Wegner, Jeffrey S Chamberlain, Rainer H A Fink, Petra Rohrbach
ZUSAMMENFASSUNG
In dystrophic mdx skeletal muscle, aberrant Ca2+ homeostasis and fibre degeneration are found. The absence of dystrophin in models of Duchenne muscular dystrophy (DMD) has been connected to altered ion channel properties e.g. impaired L-type Ca2+ currents. In regenerating mdx muscle, 'revertant' fibres restore dystrophin expression. Their functionality involving DHPR-Ca2+-channels is elusive. We developed a novel 'in-situ' confocal immuno-fluorescence and imaging technique that allows, for the first time, quantitative subcellular dystrophin-DHPR colocalization in individual, non-fixed, muscle fibres. Tubular DHPR signals alternated with second harmonic generation signals originating from myosin. Dystrophin-DHPR colocalization was substantial in wt fibres, but diminished in most mdx fibres. Mini-dystrophin (MinD) expressing fibres successfully restored colocalization. Interestingly, in some aged mdx fibres, colocalization was similar to wt fibres. Most mdx fibres showed very weak membrane dystrophin staining and were classified 'mdx-like'. Some mdx fibres, however, had strong 'wt-like' dystrophin signals and were identified as 'revertants'. Split mdx fibres were mostly 'mdx-like' and are not generally 'revertants'. Correlations between membrane dystrophin and DHPR colocalization suggest a restored putative link in 'revertants'. Using the two-micro-electrode-voltage clamp technique, Ca2+-current amplitudes (i(max)) showed very similar behaviours: reduced amplitudes in most aged mdx fibres (as seen exclusively in young mdx mice) and a few mdx fibres, most likely 'revertants', with amplitudes similar to wt or MinD fibres. Ca2+ current activation curves were similar in 'wt-like' and 'mdx-like' aged mdx fibres and are not the cause for the differences in current amplitudes. i(max) amplitudes were fully restored in MinD fibres. We present evidence for a direct/indirect DHPR-dystrophin interaction present in wt, MinD and 'revertant' mdx fibres but absent in remaining mdx fibres. Our imaging technique reliably detects single isolated 'revertant' fibres that could be used for subsequent physiological experiments to study mechanisms and therapy concepts in DMD.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin aus Rinderserum, fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, protease free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
Albumin aus Rinderserum, cold ethanol fraction, pH 5.2, ≥96%
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, protease free, pH 7, ≥98%
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, suitable for RIA, pH 5.2, ≥96%
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, lyophilized powder, essentially fatty acid free, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, low endotoxin, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, crystallized, ≥98.0% (GE)
Sigma-Aldrich
Albumin aus Rinderserum, powder, BioXtra
Sigma-Aldrich
Saponin, Molecular Biology, used as non-ionic surfactant
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, essentially globulin free, low endotoxin, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Saponin, used as non-ionic surfactant
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, suitable for (for molecular biology), Non-acetylated, Molecular Biology
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, protease free, suitable for hybridization, pH 7, ≥98%
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
Albumin aus Rinderserum, Cohn Fraction V, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, essentially fatty acid free and essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, protease free, low endotoxin, suitable for cell culture, pH 7, ≥98%
Sigma-Aldrich
Albumin aus Rinderserum, pH <5.0, powder
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, essentially globulin free, BioReagent, suitable for cell culture
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, 1× crystallized, ≥97% (agarose gel electrophoresis)