Merck
  • Membrane-bound progesterone receptors in the canine uterus and placenta; possible targets in the maintenance of pregnancy.
Membrane-bound progesterone receptors in the canine uterus and placenta; possible targets in the maintenance of pregnancy.
Theriogenology (2023-07-23)
Ali Kazemian, Miguel Tavares Pereira, Selim Aslan, Rita Payan-Carreira, Iris M Reichler, Reha A Agaoglu, Mariusz P Kowalewski
ZUSAMMENFASSUNG
To date, the biological functions of P4 within the canine placenta have been attributed to maternal stroma-derived decidual cells as the only placental cells expressing the nuclear P4 receptor (PGR). However, P4 can also exert its effects via membrane-bound receptors. To test the hypothesis that membrane-bound P4 receptors are involved in regulating placental function in the dog, the expression of mPRα, -β, -γ, PGRMC1 and -2 was investigated in the uterine and placental compartments derived from different stages of pregnancy and from prepartum luteolysis. Further, to assess the PGR signaling-mediated effects upon membrane P4 receptors in canine decidual cells, in vitro decidualized dog uterine stromal (DUS) cells were treated with type II antigestagens (aglepristone or mifepristone). The expression of all membrane P4 receptors was detectable in reproductive tissues and in DUS cells. The main findings indicate their distinguishable placental spatio-temporal distribution; PGRMC2 was predominantly found in decidual cells, PGRMC1 was strong in maternal endothelial compartments, and syncytiotrophoblast showed abundant levels of mPRα and mPRβ. In vitro decidualization was associated with increased expression of PGRMC1 and -2, while their protein levels were diminished by antigestagen treatment. The involvement of membrane-bound P4 signaling in the regulation of canine placental function is implied, with P4 effects being directly exerted through maternal and fetal cellular compartments. The indirect effects of PGR might involve the modulation of membrane-bound receptors availability in decidual cells, implying a self-regulatory loop of P4 in regulating the availability of its own receptors in the canine placenta.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N6,2′-O-Dibutyryladenosin 3′,5′-zyklisches Monophosphat Natriumsalz Monohydrat, ≥96% (HPLC), powder