Merck
Small-Molecule Aptamer for Regulating RNA Functions in Mammalian Cells and Animals.
Journal of the American Chemical Society (2023-03-31)
Keisuke Fukunaga, V Dhamodharan, Nao Miyahira, Yoko Nomura, Kamila Mustafina, Yasuaki Oosumi, Kosuke Takayama, Akira Kanai, Yohei Yokobayashi
ZUSAMMENFASSUNG
Synthetic riboswitches that can regulate gene expression by a small molecule recognized by an RNA aptamer in mammalian cells have various potential applications in biotechnology and medicine. However, the variety of small molecules and their cognate aptamers that have been demonstrated to function in mammalian cells is limited. The currently available aptamer-ligand pairs also require high small molecule concentrations to enable gene regulation, making them less desirable for industrial and biomedical applications. We conducted in vitro selection of RNA aptamers against a small molecule ASP7967 whose structure is closely related to ASP2905, a known inhibitor of potassium voltage-gated channel sub-family H member 3 (KCNH3). One of the aptamers selected (AC17-4) was found to be functional in HEK293 cells, and it was used to design aptazyme-based riboswitches that can activate gene expression (>10-fold) in the presence of ASP2905 or ASP7967 at as low as 5 μM in the culture medium. An aptazyme-based riboswitch was successfully used to regulate human erythropoietin expression in mice injected with an adeno-associated virus (AAV8) vector using orally administered ASP7967. Furthermore, by combining aptazyme-based and exon-skipping riboswitch mechanisms, an ON/OFF ratio approaching 300 was achieved with a low basal expression level in cultured cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
ASP2905, ≥98% (HPLC)
Sigma-Aldrich
ASP7967, ≥98% (HPLC)