Merck
  • Design, synthesis, and biological evaluation of 6alpha- and 6beta-N-heterocyclic substituted naltrexamine derivatives as mu opioid receptor selective antagonists.
Design, synthesis, and biological evaluation of 6alpha- and 6beta-N-heterocyclic substituted naltrexamine derivatives as mu opioid receptor selective antagonists.
Journal of medicinal chemistry (2009-02-10)
Guo Li, Lindsey C Aschenbach, Jianyang Chen, Michael P Cassidy, David L Stevens, Bichoy H Gabra, Dana E Selley, William L Dewey, Richard B Westkaemper, Yan Zhang
ZUSAMMENFASSUNG
Opioid receptor selective antagonists are important pharmacological probes in opioid receptor structural characterization and opioid agonist functional study. Thus far, a nonpeptidyl, highly selective and reversible mu opioid receptor (MOR) antagonist is unavailable. On the basis of our modeling studies, a series of novel naltrexamine derivatives have been designed and synthesized. Among them, two compounds were identified as leads based on the results of in vitro and in vivo assays. Both of them displayed high binding affinity for the MOR (K(i) = 0.37 and 0.55 nM). Compound 6 (NAP) showed over 700-fold selectivity for the MOR over the delta receptor (DOR) and more than 150-fold selectivity over the kappa receptor (KOR). Compound 9 (NAQ) showed over 200-fold selectivity for the MOR over the DOR and approximately 50-fold selectivity over the KOR. Thus these two novel ligands will serve as leads to further develop more potent and selective antagonists for the MOR.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
NAQ, ≥98% (HPLC)