Merck
  • Mapping the immune response to the outer domain of a human immunodeficiency virus-1 clade C gp120.
Mapping the immune response to the outer domain of a human immunodeficiency virus-1 clade C gp120.
The Journal of general virology (2008-09-18)
Hongying Chen, Xiaodong Xu, Hsin-Hui Lin, Ssu-Hsien Chen, Anna Forsman, Marlen Aasa-Chapman, Ian M Jones
ZUSAMMENFASSUNG
The outer domain (OD) of human immunodeficiency virus (HIV)-1 gp120 represents an attractive, if difficult, target for a beneficial immune response to HIV infection. Unlike the entire gp120, the OD is structurally stable and contains the surfaces that interact with both the primary and secondary cellular receptors. The primary strain-specific neutralizing target, the V3 loop, lies within the OD, as do epitopes for two cross-reactive neutralizing monoclonal antibodies (mAbs), b12 and 2G12, and the contact sites for a number of inhibitory lectins. The OD is poorly immunogenic, at least in the context of complete gp120, but purposeful OD immunization can lead to a substantial antibody response. Here, we map the antibody generated following immunization with a clade C OD. In contrast to published data for the clade B OD, the majority of the polyclonal response to the complete clade C OD is to the V3 loop; deletion of the loop substantially reduces immunogenicity. When the loop sequence was substituted for the epitope for 2F5, a well-characterized human cross-neutralizing mAb, a polyclonal response to the epitope was generated. A panel of mAbs against the clade C OD identified two mAbs that reacted with the loop and were neutralizing for clade C but not B isolates. Other mAbs recognized both linear and conformational epitopes in the OD. We conclude that, as for complete gp120, V3 immunodominance is a property of OD immunogens, that the responses can be neutralizing and that it could be exploited for the presentation of other epitopes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoclonal Anti-HIV-1, gp120 antibody produced in mouse, clone 1H8, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-HIV-1, gp120 antibody produced in mouse, clone 4D3, purified immunoglobulin, buffered aqueous solution