Merck
  • A new brain-penetrant glucosylceramide synthase inhibitor as potential Therapeutics for Gaucher disease.
A new brain-penetrant glucosylceramide synthase inhibitor as potential Therapeutics for Gaucher disease.
Journal of neurochemistry (2021-08-17)
Takahiro Fujii, Yuta Tanaka, Hideyuki Oki, Sho Sato, Sachio Shibata, Takamitsu Maru, Yuta Tanaka, Maiko Tanaka, Tomohiro Onishi
ZUSAMMENFASSUNG
Gaucher disease (GD), the most common lysosomal storage disorders, is caused by GBA gene mutations resulting in glycosphingolipids accumulations in various tissues, such as the brain. While suppressing glycosphingolipid accumulation is the central strategy for treating peripheral symptoms of GD, there is no effective treatment for the central nervous system symptoms. As glycosphingolipid biosynthesis starts from ceramide glycosylation by glucosylceramide synthase (GCS), inhibiting GCS in the brain is a promising strategy for neurological GD. Herein, we discovered T-036, a potent and brain-penetrant GCS inhibitor with a unique chemical structure and binding property. T-036 does not harbor an aliphatic amine moiety and has a noncompetitive inhibition mode to the substrates, unlike other known inhibitors. T-036 exhibited sufficient exposure and a significant reduction of glucosylsphingolipids in the plasma and brain of the GD mouse model. Therefore, T-036 could be a promising lead molecule for treating central nervous system symptoms of GD.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
TP-060, ≥98% (HPLC)