Merck
  • Tumor-derived interleukin-34 creates an immunosuppressive and chemoresistant tumor microenvironment by modulating myeloid-derived suppressor cells in triple-negative breast cancer.
Tumor-derived interleukin-34 creates an immunosuppressive and chemoresistant tumor microenvironment by modulating myeloid-derived suppressor cells in triple-negative breast cancer.
Cancer immunology, immunotherapy : CII (2022-09-15)
Nabeel Kajihara, Takuto Kobayashi, Ryo Otsuka, Junko Nio-Kobayashi, Tomohiro Oshino, Masato Takahashi, Seiichi Imanishi, Ari Hashimoto, Haruka Wada, Ken-Ichiro Seino
ZUSAMMENFASSUNG
Triple-negative breast cancer (TNBC) is an aggressive breast cancer subtype characterized by a lack of therapeutic targets. The paucity of effective treatment options motivated a number of studies to tackle this problem. Immunosuppressive cells infiltrated into the tumor microenvironment (TME) of TNBC are currently considered as candidates for new therapeutic targets. Myeloid-derived suppressor cells (MDSCs) have been reported to populate in the TME of TNBC, but their roles in the clinical and biological features of TNBC have not been clarified. This study identified that interleukin-34 (IL-34) released by TNBC cells is a crucial immunomodulator to regulate MDSCs accumulation in the TME. We provide evidence that IL-34 induces a differentiation of myeloid stem cells into monocytic MDSCs (M-MDSCs) that recruits regulatory T (Treg) cells, while suppressing a differentiation into polymorphonuclear MDSCs (PMN-MDSCs). As a result, the increase in M-MDSCs contributes to the creation of an immunosuppressive TME, and the decrease in PMN-MDSCs suppresses angiogenesis, leading to an acquisition of resistance to chemotherapy. Accordingly, blockade of M-MDSC differentiation with an estrogen receptor inhibitor or anti-IL-34 monoclonal antibody suppressed M-MDSCs accumulation causing retardation of tumor growth and restores chemosensitivity of the tumor by promoting PMN-MDSCs accumulation. This study demonstrates previously poorly understood mechanisms of MDSCs-mediated chemoresistance in the TME of TNBC, which is originated from the existence of IL-34, suggesting a new rationale for TNBC treatment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-IL-34, clone 1D12 Antibody, ascites fluid, clone 1D12, from mouse