Merck
Maf1 controls retinal neuron number by both RNA Pol III- and Pol II-dependent mechanisms.
iScience (2023-12-13)
Yifei Li, Dongchang Xiao, Haiqiao Chen, X F Steven Zheng, Mengqing Xiang
ZUSAMMENFASSUNG
The generation of appropriate numbers and types of neurons is a prerequisite for assembling functional neural circuits. However, the molecular basis regulating retinal neuron number remains poorly understood. Here, we report that inactivation of the RNA polymerase (Pol) III inhibitor gene Maf1 in mice results in decreased retinal thickness and neuron number that cause attenuated electroretinogram (ERG) responses. Its absence causes aberrant differentiation of all retinal neuron types primarily by an RNA Pol II-dependent mechanism while promoting retinal progenitor cell proliferation via both Pol III- and Pol II-dependent mechanisms. Chromatin profiling and transcription assay reveal that Maf1 binds widely to the genome to regulate the expression of a large set of Pol II-transcribed genes involved in retinal cell proliferation, differentiation, and/or survival. Together, our data suggest that Maf1 may control retinal neuron number by a balanced regulation of cell proliferation, differentiation, and death via both Pol III-dependent and Pol II-dependent mechanisms.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Sox9-Antikörper, Chemicon®, from rabbit
Sigma-Aldrich
Anti-GAD67-Antikörper, Klon 1G10.2, clone 1G10.2, Chemicon®, from mouse
Sigma-Aldrich
Anti-Phospho-Histon H3 (Ser10)-Antikörper, Mitosemarker, Upstate®, from rabbit
Sigma-Aldrich
Anti-Arrestin-C-Antikörper, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Glutaminsynthetase-Antikörper, Klon GS-6, clone GS-6, Chemicon®, from mouse
Sigma-Aldrich
Anti-Brn-3a-Antikörper, POU-Domäne, Protein, Klon 5A3.2, culture supernatant, clone 5A3.2, Chemicon®
Sigma-Aldrich
Anti-Protein Kinase Cα antibody produced in rabbit, whole antiserum
Sigma-Aldrich
Anti-PAX6-Antikörper, from rabbit, purified by affinity chromatography