- σE of Streptomyces coelicolor can function both as a direct activator or repressor of transcription.
σE of Streptomyces coelicolor can function both as a direct activator or repressor of transcription.
Communications biology (2024-01-07)
Jiří Pospíšil, Marek Schwarz, Alice Ziková, Dragana Vítovská, Miluše Hradilová, Michal Kolář, Alena Křenková, Martin Hubálek, Libor Krásný, Jiří Vohradský
PMID38184746
ZUSAMMENFASSUNG
σ factors are considered as positive regulators of gene expression. Here we reveal the opposite, inhibitory role of these proteins. We used a combination of molecular biology methods and computational modeling to analyze the regulatory activity of the extracytoplasmic σE factor from Streptomyces coelicolor. The direct activator/repressor function of σE was then explored by experimental analysis of selected promoter regions in vivo. Additionally, the σE interactome was defined. Taken together, the results characterize σE, its regulation, regulon, and suggest its direct inhibitory function (as a repressor) in gene expression, a phenomenon that may be common also to other σ factors and organisms.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-HA-Antikörper, monoklonaler Antikörper der Maus in Maus hergestellte Antikörper, clone HA-7, purified from hybridoma cell culture