Merck
  • Mycobacterium bovis BCG decreases MHC-II expression in vivo on murine lung macrophages and dendritic cells during aerosol infection.
Mycobacterium bovis BCG decreases MHC-II expression in vivo on murine lung macrophages and dendritic cells during aerosol infection.
Cellular immunology (2008-09-03)
Nicole D Pecora, Scott A Fulton, Scott M Reba, Michael G Drage, Daimon P Simmons, Nancy J Urankar-Nagy, W Henry Boom, Clifford V Harding
ZUSAMMENFASSUNG
Mycobacterium tuberculosis and M. bovis BCG infect APCs. In vitro, mycobacteria inhibit IFN-gamma-induced MHC-II expression by macrophages, but the effects of mycobacteria on lung APCs in vivo remain unclear. To assess MHC-II expression on APCs infected in vivo, mice were aerosol-infected with GFP-expressing BCG. At 28 d, approximately 1% of lung APCs were GFP+ by flow cytometry and CFU data. Most GFP+ cells were CD11b(high)/CD11c(neg-mid) lung macrophages (58-68%) or CD11b(high)/CD11c(high) DCs (28-31%). Lung APC MHC-II expression was higher in infected mice than naïve mice. Within infected lungs, however, MHC-II expression was lower in GFP+ cells than GFP- cells for both macrophages and DCs. MHC-II expression was also inhibited on purified lung macrophages and DCs that were infected with BCG in vitro. Thus, lung APCs that harbor mycobacteria in vivo have decreased MHC-II expression relative to uninfected APCs from the same lung, possibly contributing to evasion of T cell responses.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2,2,2-Tribromethanol, 97%