Merck
Low-dose naltrexone extends healthspan and lifespan in C. elegans via SKN-1 activation.
iScience (2024-05-27)
Weisha Li, Rebecca L McIntyre, Bauke V Schomakers, Rashmi Kamble, Anne H G Luesink, Michel van Weeghel, Riekelt H Houtkooper, Arwen W Gao, Georges E Janssens
ZUSAMMENFASSUNG
As the global aging population rises, finding effective interventions to improve aging health is crucial. Drug repurposing, utilizing existing drugs for new purposes, presents a promising strategy for rapid implementation. We explored naltrexone from the Library of Integrated Network-based Cellular Signatures (LINCS) based on several selection criteria. Low-dose naltrexone (LDN) has gained attention for treating various diseases, yet its impact on longevity remains underexplored. Our study on C. elegans demonstrated that a low dose, but not high dose, of naltrexone extended the healthspan and lifespan. This effect was mediated through SKN-1 (NRF2 in mammals) signaling, influencing innate immune gene expression and upregulating oxidative stress responses. With LDN's low side effects profile, our findings underscore its potential as a geroprotector, suggesting further exploration for promoting healthy aging in humans is warranted.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder