Merck
  • Targeting SRSF2 mutations in leukemia with RKI-1447: A strategy to impair cellular division and nuclear structure.
Targeting SRSF2 mutations in leukemia with RKI-1447: A strategy to impair cellular division and nuclear structure.
iScience (2024-04-01)
Minhua Su, Tom Fleischer, Inna Grosheva, Melanie Bokstad Horev, Malgorzata Olszewska, Camilla Ciolli Mattioli, Haim Barr, Alexander Plotnikov, Silvia Carvalho, Yoni Moskovich, Mark D Minden, Noa Chapal-Ilani, Alexander Wainstein, Eirini P Papapetrou, Nili Dezorella, Tao Cheng, Nathali Kaushansky, Benjamin Geiger, Liran I Shlush
ZUSAMMENFASSUNG
Spliceosome machinery mutations are common early mutations in myeloid malignancies; however, effective targeted therapies against them are still lacking. In the current study, we used an in vitro high-throughput drug screen among four different isogenic cell lines and identified RKI-1447, a Rho-associated protein kinase inhibitor, as selective cytotoxic effector of SRSF2 mutant cells. RKI-1447 targeted SRSF2 mutated primary human samples in xenografts models. RKI-1447 induced mitotic catastrophe and induced major reorganization of the microtubule system and severe nuclear deformation. Transmission electron microscopy and 3D light microscopy revealed that SRSF2 mutations induce deep nuclear indentation and segmentation that are apparently driven by microtubule-rich cytoplasmic intrusions, which are exacerbated by RKI-1447. The severe nuclear deformation in RKI-1447-treated SRSF2 mutant cells prevents cells from completing mitosis. These findings shed new light on the interplay between microtubules and the nucleus and offers new ways for targeting pre-leukemic SRSF2 mutant cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Proteasehemmer-Cocktail, for use with mammalian cell and tissue extracts, DMSO solution